missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Apolipoprotein C-II Protein

A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-II, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5665-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966829

  • £380.58 / 100µg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000474.2
Apolipoprotein C-II Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
344
Greater than 90% as determined by SDS-PAGE.
Research Use Only
APOC2
Recombinant Protein
E. coli
TQQPQQDEMP SPTFLTQVKE SLSSYWESAK TAAQNLYEKT YLPAVDEKLR DLYSKSTAAM STYTGIFTDQ VLSVLKGEE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers