missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOD Protein
A cDNA sequence encoding the APOD was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 347 |
---|---|
Name | APOD Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | APOD |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945988
|
enQuireBio™
QP11050-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925988
|
enQuireBio™
QP11050-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935988
|
enQuireBio™
QP11050-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
347 | |
Research Use Only | |
APOD | |
Human | |
His | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4. |
APOD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human (HEK293) | |
QAFHLGKCPN PPVQENFDVN KYLGRWYEIE KIPTTFENGR CIQANYSLME NGKIKVLNQE LRADGTVNQI EGEATPVNLT EPAKLEVKFS WFMPSAPYWI LATDYENYAL VYSCTCIIQL FHVDFAWILA RNPNLPPETV DSLKNILTSN NIDVKKMTVT DQVNCPKLSH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |