missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOBEC4 Protein
A cDNA sequence encoding the APOBEC4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | APOBEC4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985978
|
enQuireBio™
QP11048-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975978
|
enQuireBio™
QP11048-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965978
|
enQuireBio™
QP11048-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
APOBEC4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
APOBEC4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPIYEE YLANHGTIVK PYYWLSFSLD CSNCPYHIRT GEEARVSLTE FCQIFGFPYG TTFPQTKHLT FYELKTSSGS LVQKGHASSC TGNYIHPESM LFEMNGYLDS AIYNNDSIRH IILYSNNSPC NEANHCCISK MYNFLITYPG ITLSIYFSQL YHTEMDFPAS AWNREALRSL ASLWPRVVLS PISGGIWHSV LHSFISGVSG SHVFQPILTG RALADRHNAY EINAITGVKP YFTDVLLQTK RNPNTKAQEA LESYPLNNAF PGQFFQMPSG QLQPNLPPDL RAPVVFVLVP LRDLPPMHMG QNPNKPRNIV RHLNMPQMSF QETKDLGRLP TGRSVEIVEI TEQFASSKEA DEKKKKKGKK | |
Greater than 85% as determined by SDS-PAGE. |