missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AP-2 complex subunit mu Protein
A DNA sequence encoding the Homo sapiens (Human) AP-2 complex subunit mu, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5658-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001020376.1 | |
AP-2 complex subunit mu Protein | |
Research Use Only | |
AP2M1 | |
Recombinant Protein | |
Yeast | |
MIGGLFIYNH KGEVLISRVY RDDIGRNAVD AFRVNVIHAR QQVRSPVTNI ARTSFFHVKR SNIWLAAVTK QNVNAAMVFE FLYKMCDVMA AYFGKISEEN IKNNFVLIYE LLDEILDFGY PQNSETGALK TFITQQGIKS QHQTKEEQSQ ITSQVTGQIG WRREGIKYRR NELFLDVLES VNLLMSPQGQ VLSAHVSGRV VMKSYLSGMP ECKFGMNDKI VIEKQGKGTA DETSKSGKQS IAIDDCTFHQ CVRLSKFDSE RSISFIPPDG EFELMRYRTT KDIILPFRVI PLVREVGRTK LEVKVVIKSN FKPSLLAQKI EVRIPTPLNT SGVQVICMKG KAKYKASENA IVWKIKRMAG MKESQISAEI ELLPTNDKKK WARPPISMNF EVPFAPSGLK VRYLKVFEPK LNYSDHDVIK WVRYIGRSGI YETRC | |
Greater than 90% as determined by SDS-PAGE. |
1173 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |