missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AP-1 complex subunit sigma-3 Protein
A DNA sequence encoding the Homo sapiens (Human) AP-1 complex subunit sigma-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5657-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001034658.1 | |
AP-1 complex subunit sigma-3 Protein | |
Research Use Only | |
AP1S3 | |
Recombinant Protein | |
E. coli | |
MIHFILLFSR QGKLRLQKWY ITLPDKERKK ITREIVQIIL SRGHRTSSFV DWKELKLVYK RYASLYFCCA IENQDNELLT LEIVHRYVEL LDKYFGNTWP FARA | |
Greater than 90% as determined by SDS-PAGE. |
130340 | |
50 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |