missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANGPTL3 Protein
A cDNA sequence encoding the ANGPTL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11019-5ug
Additional Details : Weight : 0.01000kg
Specifications
27329 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPAECTTIYN RGEHTSGMYA IRPSNSQVFH VYCDVISGSP WTLIQHRIDG SQNFNETWEN YKYGFGRLDG EFWLGLEKIY SIVKQSNYVL RIELEDWKDN KHYIEYSFYL GNHETNYTLH LVAITGNVPN AIPENKDLVF STWDHKAKGH FNCPEGYSGG WWWHDECGEN NLNGKYNKPR AKSKPERRRG LSWKSQNGRL YSIKSTKMLI HPTDSESFE | |
Greater than 90% as determined by SDS-PAGE. |
ANGPTL3 Protein | |
Research Use Only | |
ANGPTL3 | |
Human | |
His | |
ANGPTL3 protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |