missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Amine oxidase [flavin-containing] B Protein

A DNA sequence encoding the Homo sapiens (Human) Amine oxidase [flavin-containing] B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6334-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002550

  • £613.01 / 200µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_000889.3
Amine oxidase [flavin-containing] B Protein
200 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
4129
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MAOB
Recombinant Protein
E. coli
SNKCDVVVVG GGISGMAAAK LLHDSGLNVV VLEARDRVGG RTYTLRNQKV KYVDLGGSYV GPTQNRILRL AKELGLETYK VNEVERLIHH VKGKSYPFRG PFPPVWNPIT YLDHNNFWRT MDDMGREIPS DAPWKAPLAE EWDNMTMKEL LDKLCWTESA KQLATLFVNL CVTAETHEVS ALWFLWYVKQ CGGTTRIIST TNGGQERKFV GGSGQVSERI MDLLGDRVKL ERPVIYIDQT RENVLVETLN HEMYEAKYVI SAIPPTLGMK IHFNPPLPMM RNQMITRVPL GSVIKCIVYY KEPFWRKKDY CGTMIIDGEE APVAYTLDDT KPEGNYAAIM GFILAHKARK LARLTKEERL KKLCELYAKV LGSLEALEPV HYEEKNWCEE QYSGGCYTTY FPPGILTQYG RVLRQPVDRI YFAGTETATH WSGYMEGAVE AGERAAREIL HAMGKIPEDE IWQSEPESVD VPAQPITTTF LERHLPSV
Videos
Documents