missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALDH2 Protein
A cDNA sequence encoding the ALDH2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10515-1mg
Additional Details : Weight : 0.01000kg
Specifications
217 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity was found to be > 250pmol/min/ug, and was obtained by measuring the increase of NADH in absorbance at 340 nm resulting from the reduction of NAD at pH 8.0 at 25C. | |
Human | |
Untagged | |
ALDH2 protein (1 mg/ml) contains 20mM Tris-HCl buffer, pH 7.5, 1mM DTT, 1mM EDTA and 10% Glycerol. |
ALDH2 Protein | |
Research Use Only | |
ALDH2 | |
Recombinant Protein | |
E. coli | |
MSAAATQAVP APNQQPEVFC NQIFINNEWH DAVSRKTFPT VNPSTGEVIC QVAEGDKEDV DKAVKAARAA FQLGSPWRRM DASHRGRLLNRLADLIERDR TYLAALETLD NGKPYVISYL VDLDMVLKCL RYYAGWADKY HGKTIPIDGD FFSYTRHEPV GVCGQIIPWN FPLLMQAWKL GPALATGNVV VMKVAEQTPL TALYVANLIK EAGFPPGVVN IVPGFGPTAG AAIASHEDVD KVAFTGSTEI GRVIQVAAGS SNLKRVTLEL GGKSPNIIMS DADMDWAVEQ AHFALFFNQG QCCCAGSRTF VQEDIYDEFV ERSVARAKSR VVGNPFDSKT EQGPQVDETQ FKKILGYINT GKQEGAKLLC GGGIAADRGY FIQPTVFGDV QDGMTIAKEE IFGPVMQILK FKTIEEVVGR ANNSTYGLAA AVFTKDLDKA NYLSQALQAGTVWVNCYDVF GAQSPFGGYK MSGSGRELGE YGLQAYTEVK TVTVKVPQKN S | |
Greater than 90.0% as determined by SDS-PAGE. |