missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AK4 Protein

A cDNA sequence encoding the AK4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10991-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945798

  • £2975.00 / 1mg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

205
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASKLLRAVI LGPPGSGKGT VCQRIAQNFG LQHLSSGHFL RENIKASTEV GEMAKQYIEK SLLVPDHVIT RLMMSELENR RGQHWLLDGF PRTLGQAEAL DKICEVDLVI SLNIPFETLK DRLSRRWIHP PSGRVYNLDF NPPHVHGIDD VTGEPLVQQE DDKPEAVAAR LRQYKDVAKP VIELYKSRGV LHQFSGTETN KIWPYVYTLF SNKITPIQSK EAY
Greater than 90% as determined by SDS-PAGE.
AK4 Protein
Research Use Only
AK4
Human
His
AK4 Human solution (1 mg/1ml) containing 20mM Tris-HCl pH 8.0, 2mM DTT, 0.1M NaCl, and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers