missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AICDA Protein

A cDNA sequence encoding the AICDA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10985-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985778

  • £99.15 / 5µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

57379
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MDSLLMNRRK FLYQFKNVRW AKGRRETYLC YVVKRRDSAT SFSLDFGYLR NKNGCHVELL FLRYISDWDL DPGRCYRVTW FTSWSPCYDC ARHVADFLRG NPNLSLRIFT ARLYFCEDRK AEPEGLRRLH RAGVQIAIMT FKDYFYCWNT FVENHERTFKAWEGLHENSV RLSRQLRRIL LPLYEVDDLR DAFRTLGL
Greater than 85.0% as determined by SDS-PAGE.
AICDA Protein
Research Use Only
AICDA
Human
His
The AICDA solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers