missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADP-ribosylation factor 6 Protein

A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5674-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966909

  • £1735.33 / 1mg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001654.1
ADP-ribosylation factor 6 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
382
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ARF6
Recombinant Protein
E. coli
MGKVLSKIFG NKEMRILMLG LDAAGKTTIL YKLKLGQSVT TIPTVGFNVE TVTYKNVKFN VWDVGGQDKI RPLWRHYYTG TQGLIFVVDC ADRDRIDEAR QELHRIINDR EMRDAIILIF ANKQDLPDAM KPHEIQEKLG LTRIRDRNWY VQPSCATSGD GLYEGLTWLT SNYKS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers