missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Adiponectin / Acrp30 / ADIPOQ Protein
Recombinant Protein
Brand: enQuireBio™ QP5279-25ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171271.1 | |
Adiponectin / Acrp30 / ADIPOQ Protein | |
Research Use Only | |
ADIPOQ | |
Recombinant Protein | |
E. coli | |
MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN | |
Greater than 95% as determined by SDS-PAGE. |
9370 | |
25 μg | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
gACRP (globular adiponectin) activity was confirmed by measuring inhibition of cytotoxicity of M1 cells. Typical ED50 values for our recombinant adiponectin in this assay are in the range of 3 ug/ml. | |
Human | |
Untagged | |
10 mM Tris, 0.75 mM DTT, pH 8.0 |