missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADH7 Protein

A DNA sequence encoding the Homo sapiens (Human) ADH7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5617-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936479

  • £1698.00 / 500µg

Estimated Shipment
05-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000664.2
ADH7 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
131
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADH7
Recombinant Protein
Yeast
MFAEIQIQDK DRMGTAGKVI KCKAAVLWEQ KQPFSIEEIE VAPPKTKEVR IKILATGICR TDDHVIKGTM VSKFPVIVGH EATGIVESIG EGVTTVKPGD KVIPLFLPQC RECNACRNPD GNLCIRSDIT GRGVLADGTT RFTCKGKPVH HFMNTSTFTE YTVVDESSVA KIDDAAPPEK VCLIGCGFST GYGAAVKTGK VKPGSTCVVF GLGGVGLSVI MGCKSAGASR IIGIDLNKDK FEKAMAVGAT ECISPKDSTK PISEVLSEMT GNNVGYTFEV IGHLETMIDA LASCHMNYGT SVVVGVPPSA KMLTYDPMLL FTGRTWKGCV FGGLKSRDDV PKLVTEFLAK KFDLDQLITH VLPFKKISEG FELLNSGQSI RTVLTF
Videos
SDS
Documents