missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Adenosylhomocysteinase Protein
A DNA sequence encoding the Homo sapiens (Human) Adenosylhomocysteinase, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_000678.1 |
---|---|
Gene ID (Entrez) | 191 |
Name | Adenosylhomocysteinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936559
|
enQuireBio™
QP5629-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956559
|
enQuireBio™
QP5629-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946559
|
enQuireBio™
QP5629-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15966559
|
enQuireBio™
QP5629-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15976559
|
enQuireBio™
QP5629-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15986559
|
enQuireBio™
QP5629-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000678.1 | |
Adenosylhomocysteinase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
191 | |
Research Use Only | |
AHCY | |
Recombinant Protein | |
E. coli | |
SDKLPYKVAD IGLAAWGRKA LDIAENEMPG LMRMRERYSA SKPLKGARIA GCLHMTVETA VLIETLVTLG AEVQWSSCNI FSTQDHAAAA IAKAGIPVYA WKGETDEEYL WCIEQTLYFK DGPLNMILDD GGDLTNLIHT KYPQLLPGIR GISEETTTGV HNLYKMMANG ILKVPAINVN DSVTKSKFDN LYGCRESLID GIKRATDVMI AGKVAVVAGY GDVGKGCAQA LRGFGARVII TEIDPINALQ AAMEGYEVTT MDEACQEGNI FVTTTGCIDI ILGRHFEQMK DDAIVCNIGH FDVEIDVKWL NENAVEKVNI KPQVDRYRLK NGRRIILLAE GRLVNLGCAM GHPSFVMSNS FTNQVMAQIE LWTHPDKYPV GVHFLPKKLD EAVAEAHLGK LNVKLTKLTE KQAQYLGMSC DGPFKPDHYR Y | |
Greater than 90% as determined by SDS-PAGE. |