missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADA Protein

A cDNA sequence encoding the ADA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10505-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954078

  • £99.15 / 2µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

100
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity: approximately >25 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme that deaminates 1.0 umol of adenosine to inosine per minute at pH 7.5 at 25C.
Human
His
ADA protein 0.5 mg/ml is supplied in 20mM Tris-HCL, pH 8.0, 1mM DTT and 20% Glycerol.
ADA Protein
Research Use Only
ADA
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAQTPAFDKP KVELHVHLDG SIKPETILYY GRRRGIALPA NTAEGLLNVI GMDKPLTLPD FLAKFDYYMP AIAGCREAIK RIAYEFVEMK AKEGVVYVEV RYSPHLLANS KVEPIPWNQA EGDLTPDEVV ALVGQGLQEG ERDFGVKARS ILCCMRHQPN WSPKVVELCK KYQQQTVVAI DLAGDETIPG SSLLPGHVQA YQEAVKSGIH RTVHAGEVGS AEVVKEAVDI LKTERLGHGY HTLEDQALYN RLRQENMHFE ICPWSSYLTG AWKPDTEHAV IRLKNDQANY SLNTDDPLIF KSTLDTDYQM TKRDMGFTEE EFKRLNINAA KSSFLPEDEK RELLDLLYKA YGMPPSASAG QNL
Greater than 85.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers