missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ABI3 Protein
A cDNA sequence encoding the ABI3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10928-10ug
Additional Details : Weight : 0.01000kg
Specifications
51225 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAELQQL QEFEIPTGRE ALRGNHSALL RVADYCEDNY VQATDKRKAL EETMAFTTQA LASVAYQVGN LAGHTLRMLD LQGAALRQVE ARVSTLGQMV NMHMEKVARR EIGTLATVQR LPPGQKVIAP ENLPPLTPYC RRPLNFGCLD DIGHGIKDLS TQLSRTGTLS RKSIKAPATP ASATLGRPPR IPEPVHLPVV PDGRLSAASS ASSLASAGSA EGVGGAPTPK GQAAPPAPPL PSSLDPPPPP AAVEVFQRPP TLEELSPPPP DEELPLPLDL PPPPPLDGDE LGLPPPPPGF GPDEPSWVPA SYLEKVVTLY PYTSQKDNEL SFSEGTVICV TRRYSDGWCE GVSSEGTGFF PGNYVEPSC | |
Greater than 85% as determined by SDS-PAGE. |
ABI3 Protein | |
Research Use Only | |
ABI3 | |
Human | |
His | |
The ABI3 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |