missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human A-kinase anchor protein 1
A DNA sequence encoding the Homo sapiens (Human) A-kinase anchor protein 4, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_003877.2 |
---|---|
Gene ID (Entrez) | 8852 |
Name | A-kinase anchor protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050101
|
enQuireBio™
QP7671-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070101
|
enQuireBio™
QP7671-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060101
|
enQuireBio™
QP7671-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080101
|
enQuireBio™
QP7671-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090101
|
enQuireBio™
QP7671-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000111
|
enQuireBio™
QP7671-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003877.2 | |
A-kinase anchor protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
8852 | |
Research Use Only | |
AKAP4 | |
Recombinant Protein | |
E. coli | |
QSPSAPPAKP PSTQRAVISP DGECSIDDLS FYVNRLSSLV IQMAHKEIKE KLEGKSKCLH HSICPSPGNK ERISPRTPAS KIASEMAYEA VELTAAEMRG TGEESREGGQ KSFLYSELSN KSKSGDKQMS QRESKEFADS ISKGLMVYAN QVASDMMVSL MKTLKVHSSG KPIPASVVLK RVLLRHTKEI VSDLIDSCMK NLHNITGVLM TDSDFVSAVK RNLFNQWKQN ATDIMEAMLK RLVSALIGEE KETKSQSLSY ASLKAGSHDP KCRNQSLEFS TMKAEMKERD KGKMKSDPCK SLTSAEKVGE HILKEGLTIW NQKQGNSCKV ATKACSNKDE KGEKINASTD SLAKDLIVSA LKLIQYHLTQ QTKGKDTCEE DCPGSTMGYM AQSTQYEKCG GGQSAKALSV KQLESHRAPG PSTCQKENQH LDSQKMDMSN IVLMLIQKLL NENPFKCEDP CEGENKCSEP RASKAASMSN RSDKAEEQCQ EHQELDCTSG MKQANGQFID KLVESVMKLC LIMAKYSNDG AALAELEEQA ASANKPNFRG TRCIHSGAMP QNYQDSLGHE VIVNNQCSTN SLQKQLQAVL QWIAASQFNV PMLYFMGDKD GQLEKLPQVS AKAAEKGYSV GGLLQEVMKF AKERQPDEAV GKVARKQLLD WLLANL | |
Greater than 80% as determined by SDS-PAGE. |