missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L9 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L9, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_000652.2 |
---|---|
Gene ID (Entrez) | 6133 |
Name | 60S ribosomal protein L9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064610
|
enQuireBio™
QP6622-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084610
|
enQuireBio™
QP6622-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074610
|
enQuireBio™
QP6622-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094610
|
enQuireBio™
QP6622-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004620
|
enQuireBio™
QP6622-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014620
|
enQuireBio™
QP6622-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000652.2 | |
60S ribosomal protein L9 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6133 | |
Research Use Only | |
RPL9 | |
Recombinant Protein | |
E. coli | |
MKTILSNQTV DIPENVDITL KGRTVIVKGP RGTLRRDFNH INVELSLLGK KKKRLRVDKW WGNRKELATV RTICSHVQNM IKGVTLGFRY KMRSVYAHFP INVVIQENGS LVEIRNFLGE KYIRRVRMRP GVACSVSQAQ KDELILEGND IELVSNSAAL IQQATTVKNK DIRKFLDGIY VSEKGTVQQA DE | |
Greater than 80% as determined by SDS-PAGE. |