missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L17 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L17, was expressed in the hosts and tags indicated.

£145.46 - £1688.38

Specifications

Name 60S ribosomal protein L17 Protein
Product Type Recombinant Protein
Protein Tag GST
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16094471
View Documents
enQuireBio™
QP8413-EC-10UG
10 μg £145.46
10µg View alternative products
Save Up to 
»
Estimated Shipment
10-11-2020
Please to purchase this item. Need a web account? Register with us today!
16034481
View Documents
enQuireBio™
QP8413-EC-50UG
50 μg £214.03
50µg View alternative products
Save Up to 
»
Estimated Shipment
10-11-2020
Please to purchase this item. Need a web account? Register with us today!
16084471
View Documents
enQuireBio™
QP8413-EC-100UG
100 μg £369.88
100µg View alternative products
Save Up to 
»
Estimated Shipment
10-11-2020
Please to purchase this item. Need a web account? Register with us today!
16014481
View Documents
enQuireBio™
QP8413-EC-200UG
200 μg £613.01
200µg View alternative products
Save Up to 
»
Estimated Shipment
10-11-2020
Please to purchase this item. Need a web account? Register with us today!
16024481
View Documents
enQuireBio™
QP8413-EC-500UG
500 μg £1075.37
500µg View alternative products
Save Up to 
»
Estimated Shipment
10-11-2020
Please to purchase this item. Need a web account? Register with us today!
16004481
View Documents
enQuireBio™
QP8413-EC-1MG
1 mg £1688.38
1mg View alternative products
Save Up to 
»
Estimated Shipment
10-11-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Specifications

60S ribosomal protein L17 Protein
GST
Human
Greater than 90% as determined by SDS-PAGE.
NP_000976.1
Recombinant Protein
E. coli
VRYSLDPENP TKSCKSRGSN LRVHFKNTRE TAQAIKGMHI RKATKYLKDV TLQKQCVPFR RYNGGVGRCA QAKQWGWTQG RWPKKSAEFL LHMLKNAESN AELKGLDVDS LVIEHIQVNK APKMRRRTYR AHGRINPYMS SPCHIEMILT EKEQIVPKPE EEVAQKKKIS QKKLKKQKLM ARE
6139
RPL17