missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L17 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L17, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8413-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000976.1 | |
60S ribosomal protein L17 Protein | |
Research Use Only | |
RPL17 | |
Recombinant Protein | |
E. coli | |
VRYSLDPENP TKSCKSRGSN LRVHFKNTRE TAQAIKGMHI RKATKYLKDV TLQKQCVPFR RYNGGVGRCA QAKQWGWTQG RWPKKSAEFL LHMLKNAESN AELKGLDVDS LVIEHIQVNK APKMRRRTYR AHGRINPYMS SPCHIEMILT EKEQIVPKPE EEVAQKKKIS QKKLKKQKLM ARE | |
Greater than 90% as determined by SDS-PAGE. |
6139 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |