missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 5-hydroxytryptamine receptor 3E Protein
A DNA sequence encoding the Homo sapiens(Human) 5-hydroxytryptamine receptor 3E, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9978-iv-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243542.1 | |
5-hydroxytryptamine receptor 3E Protein | |
Research Use Only | |
HTR3E | |
Recombinant Protein | |
E. coli | |
MSAILDVNEQ LHLLSSFLWL EMVWDNPFIS WNPEECEGIT KMSMAAKNLW LPDIFIIELM DVDKTPKGLT AYVSNEGRIR YKKPMKVDSI CNLDIFYFPF DQQNCTLTFS SFLYTVDSML LDMEKEVWEI TDASRNILQT HGEWELLGLS KATAKLS | |
Greater than 85% as determined by SDS-PAGE. |
285242 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |