missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S18 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8380-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_072045.1 | |
40S ribosomal protein S18 Protein | |
Research Use Only | |
RPS18 | |
Recombinant Protein | |
E. coli | |
SLVIPEKFQH ILRVLNTNID GRRKIAFAIT AIKGVGRRYA HVVLRKADID LTKRAGELTE DEVERVITIM QNPRQYKIPD WFLNRQKDVK DGKYSQVLAN GLDNKLREDL ERLKKIRAHR GLRHFWGLRV RGQHTKTTGR RGRTVGVSKK K | |
Greater than 90% as determined by SDS-PAGE. |
6222 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |