missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 39S ribosomal protein L42, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L42, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6382-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002910

  • £384.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_054769.1
39S ribosomal protein L42, mitochondrial Protein
Research Use Only
MRPL42
Recombinant Protein
E. coli
KSTYSPLPDD YNCNVELALT SDGRTIVCYH PSVDIPYEHT KPIPRPDPVH NNEETHDQVL KTRLEEKVEH LEEGPMIEQL SKMFFTTKHR WYPHGRYHRC RKNLNPPKDR
Greater than 90% as determined by SDS-PAGE.
28977
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers