missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 39S ribosomal protein L1, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L1, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8132-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002831

  • £1735.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_064621.3
39S ribosomal protein L1, mitochondrial Protein
Research Use Only
MRPL1
Recombinant Protein
E. coli
KKTKKGAKEK TPDEKKDEIE KIKAYPYMEG EPEDDVYLKR LYPRQIYEVE KAVHLLKKFQ ILDFTSPKQS VYLDLTLDMA LGKKKNVEPF TSVLSLPYPF ASEINKVAVF TENASEVKIA EENGAAFAGG TSLIQKIWDD EIVADFYVAV PEIMPELNRL RKKLNKKYPK LSRNSIGRDI PKMLELFKNG HEIKVDEERE NFLQTKIATL DMSSDQIAAN LQAVINEVCR HRPLNLGPFV VRAFLRSSTS EGLLLKIDPL LPKEVKNEES EKED
Greater than 90% as determined by SDS-PAGE.
65008
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers