missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HSV HSV-2 gD Protein

A cDNA sequence encoding the HSV-2 gD was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12343-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960119

  • £173.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSV HSV-2 gD Protein
Research Use Only
Recombinant Protein
E. coli
25mM Tris pH 7.2, 1mM EDTA, and 50% glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HSV
GST
HSV-2 gD protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers