missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV E2 Protein
A cDNA sequence encoding the HCV E2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12157-1mg
Additional Details : Weight : 0.01000kg
Specifications
HSV HCV E2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSERPY CWHYPPRPCG IVPAKSVCGP VYCFTPSPVV VGTTDRSGAP TYSWGANDTD VFVLNNTRPP LGNWFGCTWM NSTGFTKVCG APPCVIGGVG NNTLLCPTDC FRKHPEATYS RCGSGPWITP RCMVDYPYRL WHYPCTINYT IFKVRMYVGG VEHRLEAACN WTRGERCDLE DRDRSELSPL LLSTTQ | |
Greater than 80.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
The HCV E2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |