missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV Core 16.8kDa Protein
A cDNA sequence encoding the HCV Core 16.8kDa was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12140-1mg
Additional Details : Weight : 0.01000kg
Specifications
HSV HCV Core 16.8kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized from 1 mg/ml in 1xPBS pH 7.4 and 0.02% SDS. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |