missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HPV HPV 11 Protein

A cDNA sequence encoding the HPV 11 was constructed and used to recombinantly synthesize the protein.

£173.00 - £1320.00

Specifications

Name HPV HPV 11 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity HPV
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15969998
View Documents Promotion Details
enQuireBio™
QP12306-100UG
100 μg
£173.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15989998
View Documents Promotion Details
enQuireBio™
QP12306-500UG
500 μg
£660.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15979998
View Documents Promotion Details
enQuireBio™
QP12306-1MG
1 mg
£1320.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

HPV HPV 11 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HPV
GST
Recombinant HPV11 solution in PBS, 3M Urea and 100mM arginine.
Research Use Only
Recombinant Protein
E. coli
VDKLWRPSDSTVYVPPPNPVSKVVATDAYVKRTNIFYHASSSRLLAVGHPYYSIKKVNKTVVPKVSGYQYRVFKVVLPDPNKFALPDSSLFDPTTQRLVWACTGLEVGRGQPLGVGVSGHPLLNKYDDVENSGGYGGNPGQDNRVNVGMDYKQTQLCMVGCAPPLGEHWGKGTQCSNTSVQNGDCPPLELITSVIQDGDMVDTGFGAMNFADLQTNKSDVPLDICGTVCKYPDYLQMAADPYGDRLFFYLRKEQMFARHFFNRAGTVGEPVPDDLLVKGGNNRSSVASSIYVHTPSGSLVSSEAQLFNKPYWLQKAQGHNNGICWGNHLFVTVVDTTRSTNMTLCASVSKSATYTNSDYKEYMRHVEEFDLQFIFQLCSITLSAEVMAYIHTMNPSVLEDWNFGLSPPPNGTLEDTYRYVQSQAITCQKPTPEKEKQDPYKDMSFWEVNLKEKFSSELDQFPLGRKFLLQSGYRGRTSARTGIKRPAVSKPSTAPKRKRTKTKKKFGTKLSR
Protein is >90% pure as determined by 10% PAGE (coomassie staining).
Videos
SDS
Documents

Documents

Product Certifications