missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-2 Envelope Protein
A cDNA sequence encoding the HIV-2 Envelope was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12267-100ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-2 Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl and 6M urea, pH 7.2. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |