missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 Protein

A cDNA sequence encoding the HIV-1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12240-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15969798

  • £1108.23 / 1mg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
100mM NaPO4, pH 6 and 0.05% SDS.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers