missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis E HEV ORF3 Protein
A cDNA sequence encoding the HEV ORF3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12223-500ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis E HEV ORF3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-HCl, pH 8.0, 10mM B-ME and 8M urea. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis E | |
Untagged | |
HEV ORF3 protein is >95% pure as determined by 10% PAGE (coomassie staining). |