missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis D HDV Protein
A cDNA sequence encoding the HDV was constructed and used to recombinantly synthesize the protein.
£173.00 - £1100.00
Specifications
Name | Hepatitis D HDV Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis D |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959708
|
enQuireBio™
QP12212-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979708
|
enQuireBio™
QP12212-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969708
|
enQuireBio™
QP12212-1MG |
1 mg |
£1100.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis D HDV Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis D | |
Untagged | |
Protein is >95% pure as determined by 10% PAGE (Coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris pH 8 and 8M urea. |