missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis D HDV Protein
A cDNA sequence encoding the HDV was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12212-100ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis D HDV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris pH 8 and 8M urea. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis D | |
Untagged | |
Protein is >95% pure as determined by 10% PAGE (Coomassie staining). |