missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C virus genotype 1b (strain HC-J4) polyProtein

A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C virus genotype 1b (strain HC-J4) polyprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9739-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048631

  • £1698.00 / 500µg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis C virus genotype 1b (strain HC-J4) polyProtein
500 μg
Not determined.
Recombinant Protein
Yeast
STNPKPQRKT KRNTNRRPQD VKFPGGGQIV GGVYLLPRRG PRLGVRATRK ASERSQPRGR RQPIPKARRP EGRAWAQPGY PWPLYGNEGL GWAGWLLSPR GSRPSWGPTD PRRRSRNLGK VIDTLTCGFA DLMGYIPLVG APLGGAARAL AHGVRVLEDG VNYATGNLPG CSFSIF
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Partial (see sequence information for more details).
Hepatitis
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents
Certificates