missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-3b Protein
A cDNA sequence encoding the HCV NS5 Genotype-3b was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | Hepatitis C HCV NS5 Genotype-3b Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919658
|
enQuireBio™
QP12194-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939658
|
enQuireBio™
QP12194-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929658
|
enQuireBio™
QP12194-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis C HCV NS5 Genotype-3b Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV NS5 Genotype-3b protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% Glycerol. |