missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS4 a+b Rhodamine Protein
A cDNA sequence encoding the HCV NS4 a+b Rhodamine was constructed and used to recombinantly synthesize the protein.
£283.00 - £1655.00
Specifications
Name | Hepatitis C HCV NS4 a+b Rhodamine Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969608
|
enQuireBio™
QP12179-100UG |
100 μg |
£283.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989608
|
enQuireBio™
QP12179-500UG |
500 μg |
£990.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979608
|
enQuireBio™
QP12179-1MG |
1 mg |
£1655.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis C HCV NS4 a+b Rhodamine Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-Hcl pH 8.0, 8M urea and 10mM B-ME. |