missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS4 a+b Rhodamine Protein
A cDNA sequence encoding the HCV NS4 a+b Rhodamine was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12179-1mg
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV NS4 a+b Rhodamine Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-Hcl pH 8.0, 8M urea and 10mM B-ME. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |