missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Mosaic-A Protein
A cDNA sequence encoding the HCV Mosaic-A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12160-1mg
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV Mosaic-A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HCV Mosaic-A protein solution containing PBS. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |