missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-3a Protein
A cDNA sequence encoding the HCV Core Genotype-3a was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | Hepatitis C HCV Core Genotype-3a Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959528
|
enQuireBio™
QP12151-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979528
|
enQuireBio™
QP12151-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969528
|
enQuireBio™
QP12151-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis C HCV Core Genotype-3a Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV Core Genotype-3a protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 1.5M urea, 25mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% glycerol. |