missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core 24 Protein
A cDNA sequence encoding the HCV Core 24 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12143-500ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV Core 24 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-Hcl pH 8.0, 1.5M urea and 10mM b-mercaptoethanol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
HCV Core 24 protein is >95% pure as determined by 10% PAGE (coomassie staining). |