missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Combined Protein
A cDNA sequence encoding the HCV Combined was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12137-100ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV Combined Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Na-PO4, pH 8.5, 2.4mM EDTA, 5mM DTT and 0.1% SDS. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
HCV protein is >95% pure as determined by 10% PAGE (coomassie staining). |