missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBV core Protein
A cDNA sequence encoding the HBV core was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12130-500ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis B HBV core Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 1.5 mM EDTA, 1.5mM Urea and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
HBV Core protein is >90% pure as determined by 10% PAGE (Coomassie staining). |