missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis B HBV core delta Protein

A cDNA sequence encoding the HBV core delta was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12131-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959468

  • £170.83 / 100µg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis B HBV core delta Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis B
Untagged
HBV Core Delta protein is >95% pure as determined by SDS-PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
7.5mM Phosphate buffer pH 7.2, 75mM NaCl, and 50% glycerol.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers