missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg adw Protein
A cDNA sequence encoding the HBsAg adw was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12125-100ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis B HBsAg adw Protein | |
Research Use Only | |
Recombinant Protein | |
P. pastoris | |
Sterile Filtered solution containing 20mM Phosphate Buffer, 154mM sodium chloride, pH 7.1. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE and HPLC. |