missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Helicobacter Pylori CagA Pylori Protein
A cDNA sequence encoding the CagA Pylori was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11255-1mg
Additional Details : Weight : 0.01000kg
Specifications
CagA Pylori Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
CagA Pylori recombinant protein is formulated in 1xPBS pH 7.4. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Helicobacter Pylori | |
Untagged | |
Greater than 95% pure as determined by 12% PAGE (Coomassie staining). |