missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Feline UMOD Protein
A cDNA sequence encoding the UMOD was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 101084248 |
---|---|
Name | UMOD Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | UMOD |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965029
|
enQuireBio™
QP13889-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955029
|
enQuireBio™
QP13889-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945029
|
enQuireBio™
QP13889-0.1MG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
101084248 | |
Research Use Only | |
UMOD | |
Feline | |
Untagged | |
The UMOD protein was lyophilized from 0.4microm filtered solution at a concentration of 0.5 mg/ml containing deionized water. |
UMOD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Feline | |
MSVCICACGV CFFLCACVCL CTSEVRSCSE CHSNATCMED GVATTCSCQV GFTGDGLTCM DLDECAISGA HNCSEDSICV NMLGSYLCTC PDGFRLMPGL GCTDVDECAE PGLSRCHALA TCVNHKGNYS CVCPAGYWGD GWHCECSPGS CGPGLDCVPE GDTLVCVDPC QAHHILDEYW RSSEYGAGYT CDSGLSGWYR FVGQGGVRLA ETCVPVLSCN TAAPMWLNGT HPSSDEGIVS RTTCAHWRGH CCLWDAPVQV KACAGYYVYN LTAPPECYLA YCTDPSSVLG TCEECSIDED CKMDDGTWSC QCKQDFNITD LSHLERRLEC GANNIKVSLS KCQLKSLGFE KVFMYLRDSQ CSGFIERGDR DWISVVTPAR DGPCGTVMTR NETHATYSNT LYLADEIIIR DLNININFAC SYPLDMKVSL KTSLQPMVST LNISVGGTGM FTVRMALFQS PAYTQPYQGS SVTLATEAFL YVGTMLDGGD LSRFALLMTN CYATPSGNAT DPLRYFIIQD RCPRTKDSTI QVVENGESPQ GRFSVQMFRF AGNYDLVYLH CEVYLCDTVN EKCKPTCSGT RFRSGGIIDQ THVLNLGPIT RKNVQAVVSR AASSSLGFLK VWLPLLLLAS LTLMSQ | |
Greater than 95% as determined by SDS-PAGE. |