missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Feline IFNG Protein
A cDNA sequence encoding the IFNG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12377-2ug
Additional Details : Weight : 0.01000kg
Specifications
493965 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQAMFFKE IEELKGYFNA SNPDVADGGS LFVDILKNWK EESDKTIIQS QIVSFYLKMF ENLKDDDQRI QRSMDTIKED MLDKLLNTSS SKRDDFLKLI QIPVNDLQVQ RKAINELFKV MNDLSPRSNL RKRKRSQNLF RGRRASK | |
Greater than 95.0% as determined by SDS-PAGE. |
IFNG Protein | |
Research Use Only | |
IFNG | |
Feline | |
His | |
IFNG protein solution (1 mg/ml) containing PBS buffer (pH 7.4) and 10% glycerol. |