missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Equine IFN g Protein
A cDNA sequence encoding the IFN g was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10675-5ug
Additional Details : Weight : 0.01000kg
Specifications
100034181 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by an anti-viral assay using human HeLa cells infected with encephalomyocarditis (EMC) virus is less than 10.0 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. | |
Equine | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4 and 5% trehalose. |
IFN g Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
QAAFFKEIEN LKEYFNASNP DVGDGGPLFL DILKNWKEDS DKKIIQSQIV SFYFKLFENL KDNQVIQKSM DTIKEDLFVK FFNSSTSKLE DFQKLIQIPV NDLKVQRKAI SELIKVMNDL SPKANLRKRK RSQNPFRGRR ALQ | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |