missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ebola Virus Ebola Zaire VP40 Protein
A cDNA sequence encoding the Ebola Zaire VP40 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11729-1mg
Additional Details : Weight : 0.01000kg
Specifications
Ebola Zaire VP40 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ebola Zaire VP40 protein solution is supplied in Phosphate buffer, 25mM arginine and 0.02% sodium azide. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Ebola Virus | |
His | |
Greater than 95% as determined by 12% PAGE (Coomassie staining). |