missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ebola Virus Ebola Sudan Protein

A cDNA sequence encoding the Ebola Sudan Protein was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11726-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15948158

  • £1320.00 / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Ebola Sudan Protein
Research Use Only
Recombinant Protein
E. coli
Ebola Sudan protein solution is supplied in Phosphate buffer and 0.02% sodium azide.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Ebola Virus
Untagged
>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers